Potassium chloride 3 M saturated with silver chloride aqueous solution for filling electrodes

Supplier: Ricca Chemical

R5918000-1A
RC5918-32EA 178.54 CAD
RC5918-32
Potassium chloride 3 M saturated with silver chloride aqueous solution for filling electrodes
Potassium chloride
Formula: KCl
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8

Order Now

Specification Test Results

Appearance Clear, colorless, odorless

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR