Nitrogen Standard, 1 ml = 1 mg N, 1000 ppm N as Nitrate (4427 ppm NO₃⁻), Ricca Chemical Company
Supplier: Ricca Chemical
R5459000-500A R5459000-120A
RC5459-16EA 159.58 CAD
RC5459-16 RC5459-4
Nitrogen Standard, 1 ml = 1 mg N, 1000 ppm N as Nitrate (4427 ppm NO₃⁻), Ricca Chemical Company
Standards Anionic Standards Nitrate Standards
Order Now
SPECIFICATIONS
Environmentally Preferable