Manganese(II) sulfate 0,31% (w/v) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Manganese sulphate, Manganous sulphate, Manganese sulfate, Manganous sulfate

R4610000-500A
RC461016EA 74.26 CAD
RC461016
Manganese(II) sulfate 0,31% (w/v) in aqueous solution
Manganese (II) sulphate
Formula: MnO₄S
MW: 151
Density: 3.25
MDL Number: MFCD00003464
CAS Number: 7785-87-7
UN: 3077
ADR: 9 III
Merck Index: 12,05782

Order Now

Specification Test Results

Appearance Clear, light pink, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR