Iodine (iodine-iodide) 0.1563 N

Supplier: Ricca Chemical
Danger

4002-32
RC400232EA 388.68 CAD
RC400232
Iodine (iodine-iodide) 0.1563 N
Iodine (iodine-iodide)

Order Now

Specification Test Results

Appearance Clear, dark brown, Iodine odor
Assay at 20 °C (traceable to NIST SRM 136) 0.1563 ± 0.0002 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR