Ferric chloride 0,025% w/v aqueous solution

Supplier: Ricca Chemical

Synonyms: Iron(III) chloride solution

R3100000-500A R3100000-1A
RC310016EA 104.28 CAD
RC310016 RC310032
Ferric chloride 0,025% w/v aqueous solution
Iron(III) chloride solutions

APHA, for BOD.

Formula: FeCl₃·6H₂O
MW: 270.3 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0001 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1

Order Now

Specification Test Results

Appearance Clear, light amber, odorless
Identification - Chloride Passes Test
Identification - Iron (Ferric) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR