Copper(I) chloride for cyanide determination

Supplier: Ricca Chemical

R2505000-500C R2505000-1C
RC250516EA 184.86 CAD
RC250516 RC2505-32
Copper(I) chloride for cyanide determination
Copper (I) chloride
Formula: ClCu
MDL Number: MFCD00010971
CAS Number: 7758-89-6
UN: 2802
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, yellowish-green, slightly pungent

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR