Conductivity Standard, 1.413 mS/cm (1413 µmho/cm) at 25 °C, Ricca Chemical Company
Supplier: Ricca Chemical
R2236520-1A R2236520-500A R2236520-20PK
RC2236.52-32EA 127.98 CAD
RC2236.52-32 RC2236.52-16 RC2236.52-20
Conductivity Standard, 1.413 mS/cm (1413 µmho/cm) at 25 °C, Ricca Chemical Company
Standards Conductivity Standards
Order Now
SPECIFICATIONS
Environmentally Preferable