Buffer Solution B, for Low Level Sulfate Analysis (Turbidimetric Method), Ricca Chemical Company
Supplier: Ricca Chemical
R1476000-500A R1476000-1A
RC147616EA 214.88 CAD
RC147616 RC147632
Buffer Solution B, for Low Level Sulfate Analysis (Turbidimetric Method), Ricca Chemical Company
Buffers Biological Buffers
Reagent 3b (Buffer Solution B) for APHA method 4500-SO42-, E: Turbidimetric Method, for Sulfate Analysis
Order Now
SPECIFICATIONS
Environmentally Preferable