Bromophenol blue 0,4% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical

Synonyms: Bromphenol Blue Sultone Form, 3,3',5,5'-Tetrabromophenolsulfonphthalein, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein, Tetrabromophenol-sulfonephthalein

R1360000-500A
RC1360-16EA 210.14 CAD
RC1360-16
Bromophenol blue 0,4% (w/v) in aqueous solution indicator
Bromophenol blue
Formula: C₁₉H₁₀Br₄O₅S
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2

Order Now

Specification Test Results

Appearance Clear, dark reddish purple, odorless
Suitability for Use Yellow (pH 4.6)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR