Bromocresol purple 0.1% (w/v) in dilute sodium hydroxide USP Test Solution (TS)

Supplier: Ricca Chemical

R1310000-500A R1310000-120A
RC1310-16EA 134.3 CAD
RC1310-16 RC1310-4
Bromocresol purple 0.1% (w/v) in dilute sodium hydroxide USP Test Solution (TS)
Bromocresol purple
Formula: C₂₁H₁₆Br₂O₅S
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011681
CAS Number: 115-40-2

Order Now

Specification Test Results

Appearance Clear, purple, odorless
Suitability for Use Yellow (pH 6.8)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR