Human Recombinant IL-2, ACF
Supplier: STEMCELL Technologies
78145 78145.1
MSPP-78145EA 207.69 CAD
MSPP-78145 MSPP-781451006
Human Recombinant IL-2, ACF
Proteins and Peptides
Interleukin 2 (IL-2) is a monomeric cytokine that was originally identified as a T cell growth factor (Gaffen and Liu). It binds to heterotrimeric receptors consisting of CD25, CD122, and CD132. Upon binding, it activates JAK3-, STAT5-, and AKT-dependent signaling pathways, which results in cellular proliferation and survival (Ma et al.). The majority of IL-2 is secreted by activated CD4+ and CD8+ T cells, although B cells and dendritic cells were found to produce IL-2 in small amounts. IL-2 downregulates immune responses to prevent autoimmunity during thymic development, influences the development of CD4+CD25+ regulatory T cells, and affects development of follicular helper T cells. IL-2 also controls inflammation by inhibiting Th17 differentiation (Banchereau et al.). High IL-2 levels in serum are associated with progression of scleroderma, rheumatoid arthritis, and gastric and non-small cell lung cancer, though no known disease can be directly attributed to the lack or excess of IL-2 (Gaffen and Liu).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').