Anti-CD16/CD32 Rat Monoclonal Antibody [clone: 2.4G2]
Supplier: STEMCELL Technologies
60161
MSPP-60161EA 1260 CAD
MSPP-60161
Anti-CD16/CD32 Rat Monoclonal Antibody [clone: 2.4G2]
Antibodies
Rat monoclonal IgG2b antibody against mouse CD16/CD32, unconjugated.
  • Dendritic cells
  • Granulocytes
  • Subsets
  • Macrophages
  • Monocytes
  • Myeloid cells
  • Nk cells
  • T cells
  • T cells other subsets
The 2.4G2 antibody reacts with a shared epitope of Fc-gamma receptors CD16 (FcRIII) and CD32 (FcRII) in the mouse. CD16 and CD32 are low affinity receptors for the Fc portion of IgG, with CD32 having a higher affinity for IgG aggregates and immune complexes. CD16 and CD32 are expressed on B cells, monocytes, macrophages, NK cells, neutrophils, granulocytes, mast cells, andMore Product Information