Anti-Ly-49A Rat Monoclonal Antibody [clone: YE1/32.8.5]
Supplier: STEMCELL Technologies
60150
MSPP-60150EA 1290.77 CAD
MSPP-60150
Anti-Ly-49A Rat Monoclonal Antibody [clone: YE1/32.8.5]
Antibodies
Rat monoclonal IgG2a antibody against mouse Ly-49A, unconjugated.
  • Nk cells
The YE1/32.8.5 antibody reacts with Ly-49A, a member of the C-type lectin family of proteins. Ly-49A is a type II transmembrane protein expressed as a 44 kDa disulfide-linked homodimer on the surface of subsets of natural killer (NK) cells, NK-T cells, and T cells by some mammalian species, but not humans. It functions to inhibit NK cell activation by binding to certain haplotypes of MHC class I molecules on target cells, specifically H-2Dd and H2Dk, which results inMore Product Information