Anti-CD22 Mouse Monoclonal Antibody [clone: HIB22]
Supplier: STEMCELL Technologies
60083
MSPP-60083EA 393.85 CAD
MSPP-60083
Anti-CD22 Mouse Monoclonal Antibody [clone: HIB22]
Antibodies
Mouse monoclonal IgG1 antibody against human, chimpanzee CD22, unconjugated.
  • B cells
The HIB22 antibody reacts with CD22 (Siglec-2), a type I transmembrane glycoprotein expressed in the cytoplasm of pro-B and pre-B cells and on the surface of mature and activated B cells and B cell lymphomas, but not on plasma cells. CD22 functions as an inhibitory receptor for B cell receptor signaling and plays roles in modulating the activation threshold of B cells and controlling CD40 signaling. CD22 comprises two isoforms generated by alternative splicing: CD22α (More Product Information