Anti-Igg2b Isotype Control Rat Monoclonal Antibody [clone: RTK4530]

Supplier: STEMCELL Technologies
60077 60077.1
MSPP-60077EA 438.46 CAD
MSPP-60077 MSPP-600771
Anti-Igg2b Isotype Control Rat Monoclonal Antibody [clone: RTK4530]
Antibodies
Rat monoclonal IgG2b, kappa isotype control antibody, unconjugated.

The RTK4530 antibody (IgG2b, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody. Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used. If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen. The use of an appropriateMore Product Information
Order Now

SPECIFICATIONS

Antigen name Igg2b Isotype Control
Environmentally Preferable
Clonality Monoclonal
Conjugation Unconjugated
Clone RTK4530
Reactivity
Host Rat
Isotype IgG2b kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR