Anti-CD117 Rat Monoclonal Antibody [clone: ACK2]
Supplier: STEMCELL Technologies
60034
MSPP-60034EA 861.54 CAD
MSPP-60034
Anti-CD117 Rat Monoclonal Antibody [clone: ACK2]
Antibodies
Rat monoclonal IgG2b antibody against mouse CD117 (c-Kit), unconjugated.
  • Hematopoietic stem
  • Progenitor cells
The ACK2 antibody reacts with CD117 (c-Kit), an ~145 kDa type 1 transmembrane receptor for c-Kit ligand (stem cell factor/steel factor) that is broadly expressed on hematopoietic stem cells in bone marrow, including pluripotent and erythroid progenitor cells and B and T lymphocyte precursors, as well as on mast cells. CD117 belongs to the tyrosine kinase receptor family and possesses five immunoglobulin-like C2-type domains and a cytoplasmicMore Product Information