Anti-CD45 Rat Monoclonal Antibody [clone: 30-F11]
Supplier: STEMCELL Technologies
60030 60030.1
MSPP-60030EA 761.54 CAD
MSPP-60030 MSPP-600301
Anti-CD45 Rat Monoclonal Antibody [clone: 30-F11]
Antibodies
Rat monoclonal IgG2b antibody against mouse CD45, unconjugated.
  • Granulocytes
  • Subsets
  • Hematopoietic Stem
  • Progenitor cells
  • Lymphocytes
  • Myeloid cells
The 30-F11 antibody reacts with an extracellular epitope found on all isoforms and both alloantigens (CD45.1 and CD45.2) of mouse CD45, a type I transmembrane glycoprotein expressed on the surface of most hematopoietic cells except mature erythrocytes, platelets and plasma cells; expression of CD45 is lost during differentiation of these cell types. CD45 is a member of the protein tyrosine phosphataseMore Product Information