Anti-TNF-alpha Mouse Monoclonal Antibody [clone: MT15B15] (PF488P)
Supplier: STEMCELL Technologies
100-1462
MSPP-100-1462EA 926.15 CAD
MSPP-100-1462
Anti-TNF-alpha Mouse Monoclonal Antibody [clone: MT15B15] (PF488P)
Antibodies
Mouse monoclonal antibody against human TNF-alpha, PF488P-conjugated.
  • Macrophages
  • T cells
This mouse monoclonal antibody (clone MT15B15) reacts with human tumor necrosis factor alpha (TNF-α). TNF-α is a pro-inflammatory cytokine primarily produced by macrophages and T cells in response to infection and inflammation. Initially generated as a ~26 kDa transmembrane precursor, TNF-α is then processed by metalloproteinases into a ~17 kDa soluble product. Both forms of TNF-α are active in trimeric form and exert their effects via binding to either TNFR1,More Product Information