Anti-NK1.1 (CD161) Mouse Monoclonal Antibody [clone: PK136] (Biotin)
Supplier: STEMCELL Technologies
100-0463
MSPP-100-0463EA 418.46 CAD
MSPP-100-0463
Anti-NK1.1 (CD161) Mouse Monoclonal Antibody [clone: PK136] (Biotin)
Antibodies
(C3H x BALB/c) F1 hybrid monoclonal IgG2a antibody against mouse NK1.1 (CD161).
  • Nk cells
The PK136 antibody reacts with murine NK1.1 (CD161), an ~80 kDa homodimeric type 2 transmembrane glycoprotein and C-type lectin receptor expressed on NK cells, NK-T cells, and rare subsets of T cells and monocytes in select strains of mice. NK1.1 is encoded by the Klrb1b and Klrb1c genes, which specify CD161b and CD161c polypeptides, respectively. CD161b is expressed only by Swiss NIH and SJL mice, and CD161c by certain strains such as C57BL, FVB/N, and NZB (butMore Product Information