Anti-CD81 (TAPA-1) Mouse Monoclonal Antibody [clone: 5A6]
Supplier: STEMCELL Technologies
100-0209
MSPP-100-0209EA 750.77 CAD
MSPP-100-0209
Anti-CD81 (TAPA-1) Mouse Monoclonal Antibody [clone: 5A6]
Antibodies
Mouse monoclonal antibody against human, rhesus, cynomolgus CD81, unconjugated.
The 5A6 antibody reacts with an epitope located in the large extracellular loop (EC2) of human CD81 (TAPA-1). CD81 exhibits broad expression, including on the surface of B and T cells, NK cells, monocytes, dendritic cells, thymocytes, endothelial cells, and fibroblasts. It is also a marker characteristic of extracellular vesicles. CD81, a 26 kDa non-glycosylated type III transmembrane protein, belongs to the tetraspanin family. Tetraspanins contain four transmembrane domains, two extracellular loops, and shortMore Product Information