CryoStor® CS10, Cryopreservation Freeze Media, STEMCELL Technologies
Supplier: STEMCELL Technologies
07931
MSPP-07931EA 1007.69 CAD
MSPP-07931
CryoStor® CS10, Cryopreservation Freeze Media, STEMCELL Technologies
Cell Freezing Media
CryoStor® CS10 is a uniquely formulated, serum-free, animal component-free and defined cryopreservation medium containing 10% dimethyl sulfoxide (DMSO). Designed to preserve cells in low temperature environments (-80 °C to -196 °C).
  • Ready-to-use
  • Serum free, protein free
  • Animal component free
  • cGMP manufactured with USP grade/highest quality components
  • FDA master file
  • Sterility, endotoxin, and cell-based quality control testing
CryoStor® CS10 provides a safe, protective environment for cells and tissues during the freezing, storage, and thawing processes. It is recommended for the cryopreservation of hepatocytes, tissue samples, human peripheral blood, human mesenchymal stem cells, human embryonic stem (ES) and induced pluripotent stem (iPS) cells, and other extremely sensitive cell types.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Order Now
SPECIFICATIONS
Environmentally Preferable