Nessler reagent, StandARd® Ammonia Nitrogen Determination, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution

M.O.C. Category N

H261-01 MACRH261-07
MKH26101CS 907.14 CAD
MKH26101 CAMKH26107
Nessler reagent, StandARd® Ammonia Nitrogen Determination, Macron Fine Chemicals™
Nessler's reagent

For nitrogen in ammonia.

Melting Pt: –38.9 °C
Density: 13.6 g/cm³ (20 °C)
MDL Number: MFCD00049656
CAS Number: 7783-33-7
EINECS: 231-990-4
UN: 3289
ADR: 6.1 (8) II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Suitability Passes Test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR