Calcium hydroxide ≥95.0%, powder, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Slaked lime

Management of Change (MOC) category = N

4195-05
MK419505EA 600.75 CAD
MK419505
Calcium hydroxide ≥95.0%, powder, AR® ACS, Macron Fine Chemicals™
Calcium hydroxide

Powder.

Formula: Ca(OH)₂
Melting Pt: 550 °C
Density: 2.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS Number: 1305-62-0
EINECS: 215-137-3
UN: 3262
ADR: 8,III
Merck Index: 13,01675

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Ca(OH)₂) ≥95.0 %
ACS - Assay CaCO₃ ≤3 %
ACS - Chloride (Cl) ≤0.03 %
ACS - Heavy Metals (as Pb) ≤0.003 %
ACS - Insoluble in HCl ≤0.03 %
ACS - Iron (Fe) ≤0.05 %
ACS - Magnesium (Mg) ≤0.5 %
ACS - Potassium (K) ≤0.05 %
ACS - Sodium (Na) ≤0.05 %
ACS - Strontium (Sr) ≤0.05 %
ACS - Sulfur Compounds (as SO₄) ≤0.1 %
Appearance (fine, white or yellowish-white powder)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR