Masterflex® Solenoid-Operated Pinch Valves for Metric Tubing, Avantor®
Supplier: CLARK SOLUTIONS
PM-0815W 12VDC N OPEN PK-0802-NO-3 12V N OPEN PK-0802-NO-3 24V N OPEN EPK-1005-NO 24V N OPEN PK-0802-NC-3 12V N CLOSED PK-0802-NC-3 24V N CLOSED EPK-1005-NC 12V N CLOSED EPK-1005-NC 24V N CLOSED EPK-1502-NC 24V N CLOSED PM 0815W, 24VDC N OPEN EPK-1502-NO 12V N OPEN EPK-1502-NO 24V N OPEN EPK-1502-NC 12V N CLOSED
MFLX98305-00EA 282.82 CAD
MFLX98305-00 MFLX98305-02 MFLX98305-03 MFLX98305-05 MFLX98305-10 MFLX98305-11 MFLX98305-12 MFLX98305-13 MFLX98305-15 MFLX98305-01 MFLX98305-06 MFLX98305-07 MFLX98305-14
Masterflex® Solenoid-Operated Pinch Valves for Metric Tubing, Avantor®
Valves Solenoid Valves
Excellent choice for viscous fluids.
  • Specifically designed for metric sized tubing
These solenoid-operated 2- and 3-Way pinch valves are ideal for use with fluids that are difficult for traditional solenoid valves to handle, such as viscous fluids. They are ideal for on/off flow control applications and where frequent changes of wetted parts are necessary. All models are compatible with silicone, C-FLEX®, and PharMed® BPT tubing. The two-way valves are available in both normally closed and normally open configurations. The three-way valves accept twoMore Product Information