Masterflex® L/S® High-Performance Precision Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Supplier: VWR International
MFLX96420-15 MFLX96420-24 MFLX96420-35 MFLX96420-36
MFLX96420-15EA 505.6 CAD
MFLX96420-15 MFLX96420-24 MFLX96420-35 MFLX96420-36
Masterflex® L/S® High-Performance Precision Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Ultra-smooth inner surface minimizes particle entrapment.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for
More Product Information