Masterflex® L/S® Precision Pump Tubing, FDA-Compliant Viton®, Avantor®
Supplier: VWR International
MFLX96412-13 MFLX96412-14 MFLX96412-16 MFLX96412-17 MFLX96412-18 MFLX96412-25
MFLX96412-13CS 228 CAD
MFLX96412-13 MFLX96412-14 MFLX96412-16 MFLX96412-17 MFLX96412-18 MFLX96412-25
Masterflex® L/S® Precision Pump Tubing, FDA-Compliant Viton®, Avantor®
Tubing Peristaltic Pump Tubing
Good for food and lab applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly
More Product Information