Dow Corning® Pharma-65 Transfer Tubing
Supplier: Liveo
4090602 4041047 4008334 4008336 MFLX96107-09
MFLX96107-00EA 334.96 CAD
MFLX96107-00 MFLX96107-01 MFLX96107-05 MFLX96107-11 MFLX96107-09
Dow Corning® Pharma-65 Transfer Tubing
Tubing Transfer Tubing
Platinum-cured silicone tubing with a very low extractable profile.
  • Global market leader in range of formulations for most extensive chemical compatibility
Applications: Pharmaceutical, biotechnology, and ultra-pure fluid processing.
Characteristics: Platinum-cured silicone tubing with a very low extractable profile; it contains no phthalates, plasticizers, latex, or other organic additives. Remains flexible and resilient for long service life. Dow Corning maintains complete traceability from silicon to silicone tubing. Durometer shore A rating of 62.
More Product Information