Masterflex® I/P® Sanitary Fitting Precision Pump Tubing Assembly, PharMed® BPT, Avantor®
Supplier: VWR International
MFLX96102-73 MFLX96102-82 MFLX96103-73 MFLX96103-88 MFLX96102-26 MFLX96103-26 MFLX96103-82 MFLX96102-70 MFLX96102-88 MFLX96203-73
MFLX96102-73EA 385.52 CAD
MFLX96102-73 MFLX96102-82 MFLX96103-73 MFLX96103-88 MFLX96102-26 MFLX96103-26 MFLX96103-82 MFLX96102-70 MFLX96102-88 MFLX96203-73
Masterflex® I/P® Sanitary Fitting Precision Pump Tubing Assembly, PharMed® BPT, Avantor®
Tubing Peristaltic Pump Tubing
Protect your light-sensitive samples.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Applications: Cell and tissue culture work, pharmaceutical, light-sensitiveMore Product Information