Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® Fuel and Lubricant, Avantor®
Supplier: VWR International
MFLX95715-12 MFLX95715-14 MFLX95715-28 MFLX95715-40 MFLX95715-18 MFLX95715-22 MFLX95715-26 MFLX95715-38 MFLX95715-42 MFLX95715-46 MFLX95715-48 MFLX95715-34 MFLX95715-24 MFLX95715-44 MFLX95715-32 MFLX95715-36 MFLX95715-10 MFLX95715-30
MFLX95715-12EA 265.44 CAD
MFLX95715-12 MFLX95715-14 MFLX95715-28 MFLX95715-40 MFLX95715-18 MFLX95715-22 MFLX95715-26 MFLX95715-38 MFLX95715-42 MFLX95715-46 MFLX95715-48 MFLX95715-34 MFLX95715-24 MFLX95715-44 MFLX95715-32 MFLX95715-36 MFLX95715-10 MFLX95715-30
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® Fuel and Lubricant, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Resistant to ozone and UV light—won't crack, swell, or harden
  • Long service life, outstanding fatigue resistance due to flexing
To complete your Ismatec® pumping system, order three-stop color-More Product Information