Masterflex® Transfer Tubing, Ester-Based Polyurethane, Avantor®
Supplier: VWR International
MFLX95680-00 MFLX95680-02 MFLX95680-04 MFLX95680-06 MFLX95680-08 MFLX95680-14 MFLX95680-16 MFLX95680-26 MFLX95680-32 MFLX95680-42 MFLX95680-12 MFLX95680-20 MFLX95680-22 MFLX95680-36 MFLX95680-46 MFLX95680-50 MFLX95680-28
MFLX95680-00EA 124.19 CAD
MFLX95680-00 MFLX95680-02 MFLX95680-04 MFLX95680-06 MFLX95680-08 MFLX95680-14 MFLX95680-16 MFLX95680-26 MFLX95680-32 MFLX95680-42 MFLX95680-12 MFLX95680-20 MFLX95680-22 MFLX95680-36 MFLX95680-46 MFLX95680-50 MFLX95680-28
Masterflex® Transfer Tubing, Ester-Based Polyurethane, Avantor®
Tubing Transfer Tubing
Worry-free petroleum-based transfer.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
The tubes are ideal for high-purity, general laboratory, petroleum-based transfer. Characteristics include plasticizer-free, low extractables, good at low temperatures, good resistance to petroleum-based chemicals, and highMore Product Information