Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Puri-Clear™ LL, Avantor®
Supplier: VWR International
MFLX95625-48 MFLX95625-10 MFLX95625-12 MFLX95625-14 MFLX95625-16 MFLX95625-18 MFLX95625-22 MFLX95625-24 MFLX95625-26 MFLX95625-28 MFLX95625-30 MFLX95625-32 MFLX95625-34 MFLX95625-36 MFLX95625-38 MFLX95625-40 MFLX95625-42 MFLX95625-44 MFLX95625-46 MFLX95699-39
MFLX95625-48EA 91.96 CAD
MFLX95625-48 MFLX95625-10 MFLX95625-12 MFLX95625-14 MFLX95625-16 MFLX95625-18 MFLX95625-22 MFLX95625-24 MFLX95625-26 MFLX95625-28 MFLX95625-30 MFLX95625-32 MFLX95625-34 MFLX95625-36 MFLX95625-38 MFLX95625-40 MFLX95625-42 MFLX95625-44 MFLX95625-46 MFLX95699-39
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Puri-Clear™ LL, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Supple and strong tubing with high stability pumping life
  • Pthalate-free, USP Class VI PVC
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing.More Product Information