Masterflex® Transfer Tubing, DEHP-Free Flexible PVC, Avantor®
Supplier: VWR International
436901805CP 43690205CP 4360605CP 4360805CP MFLX95505-08 MFLX95505-16 MFLX95505-26 MFLX95505-28 MFLX95505-44
MFLX95505-00EA 54.35 CAD
MFLX95505-00 MFLX95505-02 MFLX95505-04 MFLX95505-06 MFLX95505-14 MFLX95505-18 MFLX95505-20 MFLX95505-22 MFLX95505-24 MFLX95505-42 MFLX95505-08 MFLX95505-16 MFLX95505-26 MFLX95505-28 MFLX95505-44
Masterflex® Transfer Tubing, DEHP-Free Flexible PVC, Avantor®
Tubing Transfer Tubing
As flexible as you need it.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Applications: Analytical instruments, general laboratory. Use for transfer of liquids, gases, and inorganic chemicals found in lab, paint, pigment, and soap dispensing applications.
Characteristics:Nontoxic DEHP-free formulation.More Product Information