Versilon™ Silver Transfter Tubing

Supplier: Saint Gobain Life Sciences
AS600027 AS600007 AS600012 AS600038 MFLX90020-03
MFLX90020-04EA 622.52 CAD
MFLX90020-04 MFLX90020-01 MFLX90020-02 MFLX90020-05 MFLX90020-03
Versilon™ Silver Transfter Tubing
Tubing Transfer Tubing
Silver ion antimicrobial coating inhibits microbe growth.

  • Global market leader in range of formulations for most extensive chemical compatibility

Formulation: Bio-based, phthalate-free flexible tubing
Applications: Food and beverage processing, dairy, laboratory, environmental, chemical, and water purification.
Characteristics: Silver ion antimicrobial coating inhibits microbe growth; not temperature sensitive. Long life, chemically resistant, nontoxic. Translucent silver.
Certifications: FDA compliant, and meets NSF-listed Standard 51.
Temperature range:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR