Dual and Quad-Head Diaphragm Pumps
Supplier: CLARK SOLUTIONS
3214-12V 3114-12V MFLX78182-00 MFLX78182-05 MFLX78182-25 MFLX78182-35 MFLX78182-45
MFLX78182-15EA 704.68 CAD
MFLX78182-15 MFLX78182-30 MFLX78182-00 MFLX78182-05 MFLX78182-25 MFLX78182-35 MFLX78182-45
Dual and Quad-Head Diaphragm Pumps
Pumps Diaphragm Pumps
6, 12, or 24 VDC driven diaphragm pumps for liquid or gas applications.
  • Run pump heads as separate entities or in series/parallel configurations
  • Oil free pumps — contamination free
  • Entry port plate can be rotated in four different positions to accommodate any application
Multiple head diaphragm pumps are an ideal option in locations where low current and high reliability are needed. Fluids do not come into contact with any metals — only silicone diaphragms and valves and polypropylene heads. Unique construction allows models 78182-40 and 78182-45 toMore Product Information