Masterflex® Ismatec® Reglo Digital Piston Pump Systems with MasterflexLive®, Avantor®
Supplier: Avantor Fluid Handling
78018-64 78018-65 78018-66 78018-61-AU 78018-62-AU 78018-64-AU 78018-65-AU 78018-66-AU 78018-61-DK 78018-62-DK 78018-64-DK 78018-65-DK 78018-66-DK 78018-61-IN 78018-62-IN 78018-64-IN 78018-65-IN 78018-66-IN 78018-61-IL 78018-62-IL 78018-64-IL 78018-65-IL 78018-66-IL 78018-61-EU 78018-62-EU 78018-64-EU 78018-65-EU 78018-66-EU 78018-61-UK 78018-62-UK 78018-64-UK 78018-65-UK 78018-66-UK 78018-61-CH 78018-62-CH 78018-64-CH 78018-65-CH 78018-66-CH 78018-61-IT 78018-62-IT 78018-64-IT 78018-65-IT 78018-66-IT 78018-63 78018-63-AU 78018-63-CH 78018-63-DK 78018-63-EU 78018-63-IN 78018-63-IT 78018-63-UK 78018-63-IL
MFLX78018-61EA 7157.4 CAD
MFLX78018-61 MFLX78018-62 MFLX78018-64 MFLX78018-65 MFLX78018-66 MFLX78018-61-AU MFLX78018-62-AU MFLX78018-64-AU MFLX78018-65-AU MFLX78018-66-AU MFLX78018-61-DK MFLX78018-62-DK MFLX78018-64-DK MFLX78018-65-DK MFLX78018-66-DK MFLX78018-61-IN MFLX78018-62-IN MFLX78018-64-IN MFLX78018-65-IN MFLX78018-66-IN MFLX78018-61-IL MFLX78018-62-IL MFLX78018-64-IL MFLX78018-65-IL MFLX78018-66-IL MFLX78018-61-EU MFLX78018-62-EU MFLX78018-64-EU MFLX78018-65-EU MFLX78018-66-EU MFLX78018-61-UK MFLX78018-62-UK MFLX78018-64-UK MFLX78018-65-UK MFLX78018-66-UK MFLX78018-61-CH MFLX78018-62-CH MFLX78018-64-CH MFLX78018-65-CH MFLX78018-66-CH MFLX78018-61-IT MFLX78018-62-IT MFLX78018-64-IT MFLX78018-65-IT MFLX78018-66-IT MFLX78018-63 MFLX78018-63-AU MFLX78018-63-CH MFLX78018-63-DK MFLX78018-63-EU MFLX78018-63-IN MFLX78018-63-IT MFLX78018-63-UK MFLX78018-63-IL
Masterflex® Ismatec® Reglo Digital Piston Pump Systems with MasterflexLive®, Avantor®
Pumps Piston Pumps
Achieve high pressure, accuracy, and repeatability—with an intuitive touch-screen interface.
  • MasterflexLive® is a secure, cloud-enabled platform for monitoring Masterflex pumps
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • 5” (12.7 cm) capacitive TFT/RGB touch screen features crisp resolution and a wide viewing angle
  • Flexible operational control via screen or analog remote control
  • Continuous run, volume dispense, and time dispense modes—batch control and interval/off time in volume and time modes
  • Pump allows for saving and
More Product Information