Masterflex® Ismatec® IPC High-Accuracy Multichannel Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
78006-22 78006-24 78006-26 78006-28 78006-42 78006-44 78006-46 78006-48 78006-22-AU 78006-24-AU 78006-26-AU 78006-28-AU 78006-42-AU 78006-44-AU 78006-46-AU 78006-48-AU 78006-22-DK 78006-24-DK 78006-26-DK 78006-28-DK 78006-42-DK 78006-44-DK 78006-46-DK 78006-48-DK 78006-22-IN 78006-24-IN 78006-26-IN 78006-28-IN 78006-42-IN 78006-44-IN 78006-46-IN 78006-48-IN 78006-22-EU 78006-24-EU 78006-26-EU 78006-28-EU 78006-42-EU 78006-44-EU 78006-46-EU 78006-48-EU 78006-22-UK 78006-24-UK 78006-26-UK 78006-28-UK 78006-42-UK 78006-44-UK 78006-46-UK 78006-48-UK 78006-22-CH 78006-24-CH 78006-26-CH 78006-28-CH 78006-42-CH 78006-44-CH 78006-46-CH 78006-48-CH 78006-22-IT 78006-24-IT 78006-26-IT 78006-28-IT 78006-42-IT 78006-44-IT 78006-46-IT 78006-48-IT 78006-22-IL 78006-24-IL 78006-26-IL 78006-28-IL 78006-42-IL 78006-44-IL 78006-46-IL 78006-48-IL
MFLX78006-22EA 10949.4 CAD
MFLX78006-22 MFLX78006-24 MFLX78006-26 MFLX78006-28 MFLX78006-42 MFLX78006-44 MFLX78006-46 MFLX78006-48 MFLX78006-22-AU MFLX78006-24-AU MFLX78006-26-AU MFLX78006-28-AU MFLX78006-42-AU MFLX78006-44-AU MFLX78006-46-AU MFLX78006-48-AU MFLX78006-22-DK MFLX78006-24-DK MFLX78006-26-DK MFLX78006-28-DK MFLX78006-42-DK MFLX78006-44-DK MFLX78006-46-DK MFLX78006-48-DK MFLX78006-22-IN MFLX78006-24-IN MFLX78006-26-IN MFLX78006-28-IN MFLX78006-42-IN MFLX78006-44-IN MFLX78006-46-IN MFLX78006-48-IN MFLX78006-22-USS MFLX78006-24-USS MFLX78006-26-USS MFLX78006-28-USS MFLX78006-42-USS MFLX78006-44-USS MFLX78006-46-USS MFLX78006-48-USS MFLX78006-22-EU MFLX78006-24-EU MFLX78006-26-EU MFLX78006-28-EU MFLX78006-42-EU MFLX78006-44-EU MFLX78006-46-EU MFLX78006-48-EU MFLX78006-22-UK MFLX78006-24-UK MFLX78006-26-UK MFLX78006-28-UK MFLX78006-42-UK MFLX78006-44-UK MFLX78006-46-UK MFLX78006-48-UK MFLX78006-22-CH MFLX78006-24-CH MFLX78006-26-CH MFLX78006-28-CH MFLX78006-42-CH MFLX78006-44-CH MFLX78006-46-CH MFLX78006-48-CH MFLX78006-22-IT MFLX78006-24-IT MFLX78006-26-IT MFLX78006-28-IT MFLX78006-42-IT MFLX78006-44-IT MFLX78006-46-IT MFLX78006-48-IT MFLX78006-22-USN MFLX78006-24-USN MFLX78006-26-USN MFLX78006-28-USN MFLX78006-42-USN MFLX78006-44-USN MFLX78006-46-USN MFLX78006-48-USN MFLX78006-22-IL MFLX78006-24-IL MFLX78006-26-IL MFLX78006-28-IL MFLX78006-42-IL MFLX78006-44-IL MFLX78006-46-IL MFLX78006-48-IL
Masterflex® Ismatec® IPC High-Accuracy Multichannel Peristaltic Pumps, Avantor®
Pumps Peristaltic Pumps
Achieve high-accuracy multi-channel flow, with an intuitive touch-screen interface.
  • Replicate same flow for all channels; perfect for auto-analysis, reactor studies, cell line seeding, and research
  • Planetary drive with 8 stainless steel rollers reduces pulsation, increases flow rate accuracy, and lengthens tubing life
  • 12.7 cm (5") capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or remote control (analog remote control, serial, or EtherNet/IP)
  • Three-level user access – gives you the
More Product Information