Masterflex® MasterSense™ Gear Pump Systems
Supplier: Avantor Fluid Handling
77976-50 77976-51 77976-52 77976-50-AU 77976-52-AU 77976-51-AU 77976-50-DK 77976-52-DK 77976-51-DK 77976-50-IN 77976-52-IN 77976-51-IN 77976-50-EU 77976-52-EU 77976-51-EU 77976-50-UK 77976-52-UK 77976-51-UK 77976-50-CH 77976-52-CH 77976-51-CH 77976-50-IT 77976-52-IT 77976-51-IT 77976-50-IL 77976-52-IL 77976-51-IL
MFLX77976-50EA 13619.6 CAD
MFLX77976-50 MFLX77976-51 MFLX77976-52 MFLX77976-50-AU MFLX77976-52-AU MFLX77976-51-AU MFLX77976-50-DK MFLX77976-52-DK MFLX77976-51-DK MFLX77976-50-IN MFLX77976-52-IN MFLX77976-51-IN MFLX77976-50-EU MFLX77976-52-EU MFLX77976-51-EU MFLX77976-50-UK MFLX77976-52-UK MFLX77976-51-UK MFLX77976-50-CH MFLX77976-52-CH MFLX77976-51-CH MFLX77976-50-IT MFLX77976-52-IT MFLX77976-51-IT MFLX77976-50-IL MFLX77976-52-IL MFLX77976-51-IL
Masterflex® MasterSense™ Gear Pump Systems
Pumps Gear Pumps
Versatile gear pump systems for high-accuracy fluid delivery - with an intuitive touch-screen interface.
  • Powerful, space-efficient drives accept all Micropump A and B mount gear pump heads
  • Pulseless gear flow technology allows for high repeatability over long duration
  • High viscosity capability (0.2 to 1500 centipoise)
  • 12.7 cm (5") capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Single-point pressure monitoring provides real-time pressure display and alarms for process reliability
  • Continuous run, volume dispense, and time
More Product Information