Masterflex® I/P® Brushless Process Drive Systems, Avantor®
Supplier: Avantor Fluid Handling
77965-00 77965-00-AU 77965-00-DK 77965-00-IN 77965-00-EU 77965-00-UK 77965-00-CH 77965-00-IT 77965-00-IL
MFLX77965-00EA 10459.6 CAD
MFLX77965-00 MFLX77965-10 MFLX77965-00-AU MFLX77965-10-AU MFLX77965-00-DK MFLX77965-10-DK MFLX77965-00-IN MFLX77965-10-IN MFLX77965-10-USS MFLX77965-00-EU MFLX77965-10-EU MFLX77965-00-UK MFLX77965-10-UK MFLX77965-00-CH MFLX77965-10-CH MFLX77965-00-IT MFLX77965-10-IT MFLX77965-10-USN MFLX77965-00-IL MFLX77965-10-IL
Masterflex® I/P® Brushless Process Drive Systems, Avantor®
Pumps Peristaltic Pumps
Powerful, low-maintenance drives are versatile and cost-effective.
  • Flow rate 0.2 to 26 LPM with I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with LED display shows percent speed with 1% resolution
  • Speed control via three-turn potentiometer for quick, easy adjustments
  • Easily control start/stop and reverse features via sealed keypad
  • Contructed of non-chip painted steel and aluminum IP55-rated housing
  • Brushless, maintenance-free motor offers ±0.25% speed control accuracy
  • Remote control available using analog
More Product Information