Masterflex® L/S® Network-Compatible Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
MFLX77924-70EA 9195.6 CAD
MFLX77924-70 MFLX77924-82 MFLX77975-60 MFLX77975-62 MFLX77975-65 MFLX77975-77 MFLX77924-70-AU MFLX77924-82-AU MFLX77975-60-AU MFLX77975-62-AU MFLX77975-65-AU MFLX77975-77-AU MFLX77924-70-DK MFLX77924-82-DK MFLX77975-60-DK MFLX77975-62-DK MFLX77975-65-DK MFLX77975-77-DK MFLX77924-70-IN MFLX77924-82-IN MFLX77975-60-IN MFLX77975-62-IN MFLX77975-65-IN MFLX77975-77-IN MFLX77924-70-IL MFLX77924-82-IL MFLX77975-60-IL MFLX77975-62-IL MFLX77975-65-IL MFLX77975-77-IL MFLX77924-70-EU MFLX77924-82-EU MFLX77975-60-EU MFLX77975-62-EU MFLX77975-65-EU MFLX77975-77-EU MFLX77924-70-UK MFLX77924-82-UK MFLX77975-60-UK MFLX77975-62-UK MFLX77975-65-UK MFLX77975-77-UK MFLX77924-70-CH MFLX77924-82-CH MFLX77975-60-CH MFLX77975-62-CH MFLX77975-65-CH MFLX77975-77-CH MFLX77924-70-IT MFLX77924-82-IT MFLX77975-60-IT MFLX77975-62-IT MFLX77975-65-IT MFLX77975-77-IT MFLX77924-80 MFLX77975-72 MFLX77975-67 MFLX77975-70 MFLX77975-75 MFLX77924-72-AU MFLX77924-80-AU MFLX77975-72-AU MFLX77924-72-DK MFLX77924-80-DK MFLX77975-72-DK MFLX77924-72-IN MFLX77924-80-IN MFLX77975-72-IN MFLX77924-72-IL MFLX77924-80-IL MFLX77975-72-IL MFLX77924-72-EU MFLX77924-80-EU MFLX77975-72-EU MFLX77924-72-UK MFLX77924-80-UK MFLX77975-72-UK MFLX77924-72-CH MFLX77924-80-CH MFLX77975-72-CH MFLX77924-72-IT MFLX77924-80-IT MFLX77975-72-IT
Masterflex® L/S® Network-Compatible Pump Systems, Avantor®
Pumps Peristaltic Pumps
Networking drives offer either native EtherNet/IP™ or Profibus communication—no additional hardware or firmware required.
  • Flow rate 0.001 to 3400 ml/min with L/S tubing—flow rate depends on drive rpm pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—
More Product Information