Masterflex® B/T® Fixed Speed Analog Drives, Avantor®
Supplier: Avantor
MFLX77111-30EA 11502.4 CAD
MFLX77111-30 MFLX77111-37
Masterflex® B/T® Fixed Speed Analog Drives, Avantor®
Pumps Peristaltic Pumps
High-volume transfer and dispensing.
  • Flow rate 18.9/16 or 42/35 LPM with B/T tubing—flow rate depends on drive and tubing size
  • Rapid-Load® pump head occlusion mechanism slides occlusion bed out of the way for easy tube loading and improved tubing occlusion
  • Accepts B/T PerfectPosition™ pump tubing sizes B/T 87 and B/T 91
  • Interlock switch shuts off power to stop drive when door is opened
Masterflex® B/T® drives are the best choice for high-flow bulk-transfer applications. Their durable construction ensures long service life with minimal maintenance.More Product Information