Masterflex® Single-Syringe Infusion Pumps, Avantor®
Supplier: VWR International
MFLX74900-00 MFLX74900-05
MFLX74900-00EA 3160 CAD
MFLX74900-00 MFLX74900-05
Masterflex® Single-Syringe Infusion Pumps, Avantor®
Pumps Syringe Pumps
Ideal for low-flow, low-pressure applications where precision is needed.
  • Flow rate of 0.0033 to 8.33 ml/min per syringe
  • Backlit alphanumeric LCD guides you through setup and operating procedures
  • Nonvolatile memory retains settings even after your pump is turned off
  • Accurate to +/-0.5%
This features microprocessor motor control and precision gearing. Microprocessor automatically selects display units based on syringe used. Obtain precise flow rates down to 0.2 µL/hr. A wide dynamic range (6000:1) provides excellent flexibility. All comm