Pump Flow Calibration Columns, PVC 40
Supplier: KOFLO CORPORATION
250 ML 100 ML 500 ML 1000 ML 2000 ML 4000 ML 10000 ML MFLX74600-08
MFLX74600-00EA 129.24 CAD
MFLX74600-00 MFLX74600-01 MFLX74600-02 MFLX74600-04 MFLX74600-05 MFLX74600-06 MFLX74600-07 MFLX74600-08
Pump Flow Calibration Columns, PVC 40
Flow Meters
Calibration columns provide a simple, economical method for checking pump flow rates.
  • Constructed of durable PVC
Columns are graduated in both ml/min and GPH to help improve system accuracy. For maximum convenience and accuracy, choose small capacity columns to check low flow rates; select larger capacity columns for higher flow rates.
To check your flow rate, install column on the suction end of your pump. Turn the pump on, and fill the column to zero (at the top). Using a ball valve, shut off supply from the storage tank and monitor time as fluid isMore Product Information