LMI Microprocessor-Controlled Solenoid Diaphragm Metering Pumps
Supplier: EQUIP SOLUTIONS
AD211-918SI AD241-928SI AD251-938SI AD811-918SI AD851-938SI AD861-948SI AD941-928SI AD961-948SI MFLX74522-08 MFLX74522-14 MFLX74522-16 MFLX74522-24 MFLX74522-42 MFLX74522-46
MFLX74522-02EA 2180.4 CAD
MFLX74522-02 MFLX74522-04 MFLX74522-06 MFLX74522-22 MFLX74522-26 MFLX74522-28 MFLX74522-44 MFLX74522-48 MFLX74522-08 MFLX74522-14 MFLX74522-16 MFLX74522-24 MFLX74522-42 MFLX74522-46
LMI Microprocessor-Controlled Solenoid Diaphragm Metering Pumps
Pumps Diaphragm Pumps
Robust processor-based control offers improved electromagnetics.
  • Easy-to-use push button controls with LCD user interface
  • Manual stroke length knob
  • Low level float switch connection
  • Lockable clear cover
  • NEMA 4X / IP65 enclosure
Choose from manual control, remote control, and digital remote control pump options.
Manual control models feature manual stroke length and frequency adjustment. Low level indication is possible via the remote input. LCD provides quick user feedback of pump status and theoretical pump flow.
Remote control models accept a pulse orMore Product Information