Masterflex® 316 Stainless Steel Syringes, Avantor®
Supplier: VWR International
MFLX74044-00 MFLX74044-12 MFLX74044-20 MFLX74044-24 MFLX74044-26 MFLX74044-32 MFLX74044-34 MFLX74044-36 MFLX74044-44 MFLX74044-46 MFLX74044-54 MFLX74044-10 MFLX74044-22 MFLX74044-30 MFLX74044-40 MFLX74044-42 MFLX74044-50 MFLX74044-52 MFLX74044-56
MFLX74044-00EA 2006.6 CAD
MFLX74044-00 MFLX74044-12 MFLX74044-20 MFLX74044-24 MFLX74044-26 MFLX74044-32 MFLX74044-34 MFLX74044-36 MFLX74044-44 MFLX74044-46 MFLX74044-54 MFLX74044-10 MFLX74044-22 MFLX74044-30 MFLX74044-40 MFLX74044-42 MFLX74044-50 MFLX74044-52 MFLX74044-56
Masterflex® 316 Stainless Steel Syringes, Avantor®
Syringes General-Purpose Syringes
Ideal for use in Masterflex® syringe pumps.
  • Syringes fit most syringe pumps
  • Made from top quality 316 stainless steel — provides superior chemical resistance
  • Designed to handle demanding applications with pressures
  • Leak free to pressures up to 7000 psi
  • Autoclavable
These Masterflex® stainless steel syringes are durable and can stand up to pressure. Stainless steel offers good resistance to most aggressive liquids. The syringe barrel and end plungers are made of 316 stainless steel. Both syringes barrel end plungers are #316 stainless steel. A Viton®More Product Information