Masterflex® Motorized Drum Pump Systems, Avantor®
Supplier: VWR International
9400-110V 9401-230V 9430-110V 9420-110V 9421-230V 9500-110V 9511-230 9710-110V
MFLX70614-02EA 3081 CAD
MFLX70614-02 MFLX70614-03 MFLX70614-00 MFLX70614-04 MFLX70614-05 MFLX70614-06 MFLX70614-09 MFLX70614-10
Masterflex® Motorized Drum Pump Systems, Avantor®
Pumps Drum/Transfer Pumps
Complete drum pump system specifically packaged for different applications.
  • Flow rate 35 GMP for all models
  • Equipped with variable-speed control motors for added safety and control
  • Maintenance is reduced with no mechanical seals
  • Run dry capability
Pre-configured drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks, and small storage vessels. Based upon the application, polypropylene (PP), CPVC, PVDF, and 316 stainless steel (SS) materials provide excellent chemical resistance against corrosive liquids.
TheMore Product Information