Flowline Switch-Tek Vibration Level Switch
Supplier: FLOWLINE
LZ12-1405
MFLX68869-04EA 960.64 CAD
MFLX68869-04
Flowline Switch-Tek Vibration Level Switch
Level Sensors
Don’t worry about buildup—automatically adjusts for optimal performance, alerts when cleaning is required.
  • Constructed of Glass filled Ryton® and Viton®
SwitchTek vibration level switch is an excellent choice for more challenging application liquids, including those with light to medium coating or scaling-type characteristics. Liquid examples include light sodium hydroxide, brine and copper sulfate solutions.