Masterflex® Direct-Reading Variable-Area Flowmeters for Water, Acrylic, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
ACRYB0V16 ACRZS0V02 ACRZS0V09 ACRZS0V16 ACRYS0V23 ACRZB1V02 ACRZB1V09 ACRZB1V16 ACRYB1V09 ACRYB1V16 ACRYB1V23 ACRZS1V02 ACRZS1V16 ACRYS1V02 ACRYS1V09 ACRYS1V16 ACRYS1V23 MFLX68561-70 MFLX68561-71 MFLX68561-72 MFLX68561-73 MFLX68561-74 MFLX68561-76 MFLX68561-80 MFLX68561-81 MFLX68561-82 MFLX68561-87 MFLX68561-92
MFLX68561-75EA 106.02 CAD
MFLX68561-75 MFLX68561-77 MFLX68561-78 MFLX68561-79 MFLX68561-83 MFLX68561-84 MFLX68561-85 MFLX68561-86 MFLX68561-88 MFLX68561-89 MFLX68561-90 MFLX68561-91 MFLX68561-93 MFLX68561-94 MFLX68561-95 MFLX68561-96 MFLX68561-97 MFLX68561-70 MFLX68561-71 MFLX68561-72 MFLX68561-73 MFLX68561-74 MFLX68561-76 MFLX68561-80 MFLX68561-81 MFLX68561-82 MFLX68561-87 MFLX68561-92
Masterflex® Direct-Reading Variable-Area Flowmeters for Water, Acrylic, Avantor®
Flow Meters
Designed for labs and plant processes where multiple gases are used.
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
Versatile acrylic flowmeters are designed specifically for water. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. FlowmetersMore Product Information