Masterflex® Direct-Reading Variable-Area Flowmeters for Carbon Dioxide, Acrylic, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
ACRYB0V04 ACRYB0V11 ACRYB0V18 ACRZS0V04 ACRYS0V11 ACRZB1V04 ACRZB1V11 ACRZB1V18 ACRYB1V04 ACRYB1V11 ACRZS1V04 ACRZS1V11 ACRYS1V04 ACRYS1V11 ACRYS1V18 ACRYS1V25 MFLX68560-84 MFLX68560-85 MFLX68560-86 MFLX68560-92 MFLX68560-93 MFLX68560-94 MFLX68560-96 MFLX68560-97 MFLX68561-05 MFLX68561-06 MFLX68561-09
MFLX68560-87EA 106.02 CAD
MFLX68560-87 MFLX68560-88 MFLX68560-89 MFLX68560-91 MFLX68560-95 MFLX68561-00 MFLX68561-01 MFLX68561-02 MFLX68561-03 MFLX68561-04 MFLX68561-07 MFLX68561-08 MFLX68561-10 MFLX68561-11 MFLX68561-12 MFLX68561-13 MFLX68560-84 MFLX68560-85 MFLX68560-86 MFLX68560-92 MFLX68560-93 MFLX68560-94 MFLX68560-96 MFLX68560-97 MFLX68561-05 MFLX68561-06 MFLX68561-09
Masterflex® Direct-Reading Variable-Area Flowmeters for Carbon Dioxide, Acrylic, Avantor®
Flow Meters
Designed for labs and plant processes where multiple gases are used.
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
Versatile acrylic flowmeters are designed specifically for carbon dioxide. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations.More Product Information