Value Plastics® Adapter Fittings, Female Luer to Hose Barb, Straight, Polycarbonate
Supplier: VALUE PLASTICS TE
FTLL210-91000 FTLL004-91000 FTLL220-91000 FTLL230-91000 FTLL240-91000 FTLL250-91000 FTLL055-91000 FTLLB210-91000
MFLX45501-00BLKEA 466.1 CAD
MFLX45501-00BLK MFLX45501-01BLK MFLX45501-02BLK MFLX45501-04BLK MFLX45501-06BLK MFLX45501-08BLK MFLX45501-20BLK MFLX45501-30BLK
Value Plastics® Adapter Fittings, Female Luer to Hose Barb, Straight, Polycarbonate
Fittings and Connectors
Ensure your process integrity with full material traceability.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly
More Product Information