Masterflex® Union Fittings, Hose Barb, Y-Connector, Polyproylene
Supplier: VWR International
MFLX06295-10 MFLX06295-20 MFLX06295-30 MFLX06295-40 MFLX06295-50 MFLX06295-60 MFLX06295-70 MFLX06296-00 MFLX06296-10 MFLX06296-20 MFLX06296-40 MFLX06296-50 MFLX06368-18 MFLX06368-19 MFLX06368-22 MFLX06368-24 MFLX06368-25 MFLX30614-43 MFLX30614-51 MFLX30614-55 MFLX30614-59 MFLX30614-63 MFLX30614-67 MFLX30614-71 MFLX40726-41 MFLX40726-42 MFLX40726-43 MFLX40726-44 MFLX40726-45 MFLX40726-46 MFLX40726-47 MFLX40726-48 MFLX40726-49 MFLX40726-50 MFLX40727-41 MFLX40727-42 MFLX40727-43 MFLX40727-44 MFLX40727-45 MFLX40727-46 MFLX41526-28 MFLX41526-39 MFLX40727-47 MFLX40617-76 MFLX40617-77 MFLX40617-79 MFLX40617-80 MFLX40728-43
MFLX06295-10EA 78.68 CAD
MFLX06295-10 MFLX06295-20 MFLX06295-30 MFLX06295-40 MFLX06295-50 MFLX06295-60 MFLX06295-70 MFLX06296-00 MFLX06296-10 MFLX06296-20 MFLX06296-40 MFLX06296-50 MFLX06368-18 MFLX06368-19 MFLX06368-22 MFLX06368-24 MFLX06368-25 MFLX30614-43 MFLX30614-51 MFLX30614-55 MFLX30614-59 MFLX30614-63 MFLX30614-67 MFLX30614-71 MFLX40726-41 MFLX40726-42 MFLX40726-43 MFLX40726-44 MFLX40726-45 MFLX40726-46 MFLX40726-47 MFLX40726-48 MFLX40726-49 MFLX40726-50 MFLX40727-41 MFLX40727-42 MFLX40727-43 MFLX40727-44 MFLX40727-45 MFLX40727-46 MFLX41526-28 MFLX41526-39 MFLX40727-47 MFLX40617-76 MFLX40617-77 MFLX40617-79 MFLX40617-80 MFLX40617-87 MFLX40728-43
Masterflex® Union Fittings, Hose Barb, Y-Connector, Polyproylene
Fittings and Connectors
No tools needed for assembly.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and
More Product Information